Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cell Separation Magnet

Catalog No. bdb552311 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
BDB552311 Each
1 options
The BD IMagnet™ contains a strong permanent magnet that may affect or damage delicate instruments, electronic equipment, and magnetic recording devices. Therefore, persons wearing cardiac pacemakers should not handle the BD IMagnet, and the BD IMagnet should not be placed near laboratory instruments, monitors, computer discs, credit cards, video or audio cassettes, and watches. The BD IMagnet has a very strong attraction for ferric metals, so it should not be placed on steel surfaces or near objects containing steel or iron.

Catalog No. BDB552311

Supplier: BD 552311

Only null left
Add to Cart
Add to Cart
The BD IMagnet™ contains a strong permanent magnet that may affect or damage delicate instruments, electronic equipment, and magnetic recording devices. Therefore, persons wearing cardiac pacemakers should not handle the BD IMagnet, and the BD IMagnet should not be placed near laboratory instruments, monitors, computer discs, credit cards, video or audio cassettes, and watches. The BD IMagnet has a very strong attraction for ferric metals, so it should not be placed on steel surfaces or near objects containing steel or iron.

Specialized plastic test-tube rack surrounding a strong permanent rare earth magnet

  • Holds up to six 12 x 75mm or two 17 x 100mm round-bottom test tubes
  • Allows rapid and effective positive or negative selection of desired leukocyte populations

Cell separation

TRUSTED_SUSTAINABILITY

Specifications

Quantity Each
For Use With (Application) Cell separation
Type Cell Separation Magnet
Sufficient For Six 12 x 75mm/two 17 x 100mm round-botto m test tubes

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.