Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cell Viability Solution

Catalog No. BDB555815 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
500 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555815 500 Tests
BDB555816 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB555815 Supplier BD Supplier No. 555815
Add to Cart
Add to Cart

Ready-to-use solution of the nucleic acid dye, 7-Amino-actinomycin D (7-AAD) that can be used in place of propidium iodide (PI) for the exclusion of nonviable cells in flow cytometric assays

  • Used as a viability probe for methods of dead cell exclusion, based on a light scatter and uptake of 7-AAD as detected in FL3
  • Does not require dilution

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
For Use With (Application) Flow Cytometry
Quantity 500 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.