Learn More
BD Cell Viability Solution
Catalog No. BDB555816 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
500 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555816 100 Tests
BDB555815 500 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB555816 Supplier BD Supplier No. 555816
Add to Cart
Add to Cart
Ready-to-use solution of the nucleic acid dye, 7-Amino-actinomycin D (7-AAD) that can be used in place of propidium iodide (PI) for the exclusion of nonviable cells in flow cytometric assays
  • Used as a viability probe for methods of dead cell exclusion, based on a light scatter and uptake of 7-AAD as detected in FL3
  • Does not require dilution
Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 4°C
Quantity 100 Tests