Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD CultureSwab™ MaxV and MaxV+ Collection and Transport Systems

Catalog No. p-3107485 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Media Type:
MaxV Liquid Amies
Swab Configuration:
Dual Applicator
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type Swab Configuration
L220232 MaxV Liquid Amies Dual Applicator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L220232 Supplier BD Supplier No. 220232
This item is not returnable. View return policy

Improved recovery of fastidious organisms for proper patient diagnosis

  • Improved recovery of fastidious organisms such as N. gonorrhoeae, H. influenzae, N. meningitidis, and S. pneumoniae
  • Sterile, ready-to-use systems intended for collection, transport, and preservation of clinical specimens for bacteriological examination
  • Medium is designed to maintain viability of organisms during transit to the laboratory
  • Pouches are made of a plastic film which retards penetration of atmospheric air into the product
Swabs contain unique blend of non-animal proteins embedded into swab fibers, to enhance recovery, in addition to all performance enhancing features of the BD CultureSwab line:
  • Nitrogen-flushed packaging
  • Patented tube design
  • Tamper-evident seal
  • Double-action security cap
  • Shaft: 5 1/4 in.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Collection and Transport System
Media Type MaxV Liquid Amies
Sterility Sterile
Swab Configuration Dual Applicator
For Use With (Application) Collection, transport and preservation of clinical specimens for Bacteriological examination
Packaging Nitrogen flushed
Quantity 50/Pk.
Storage Requirements 5°C to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.