Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human Anaphylatoxin Kit

Catalog No. bdb561418 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
Catalog No. Quantity
BDB561418 25 Tests
1 options

Catalog No. BDB561418

Supplier: BD 561418

Only null left
Add to Cart
Add to Cart

Can be used to quantitatively measure anaphylatoxin C3a, C4a, and C5a (bioactive cleavage products released from C3, C4, and C5 during complement activation) protein levels in a single EDTA plasma or serum sample.

  • Performance has been optimized for analysis of specific proteins in EDTA plasma and serum samples
  • Provides sufficient reagents for the quantitative analysis of 80 samples

Applications:

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Anaphylatoxin
Cross Reactivity Human
Includes Reagents for capture and detection of C4a, C3a and C5a, standards, Wash Buffer, assay diluent, cytometer setup reagents
Quantity 25 Tests
Content And Storage 2°C to 8°C
For Use With (Application) Flow Cytometry
Product Type Human anaphylatoxin kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.