Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human Enhanced Sensitivity Detection Reagent (Part B)
Shop All BD Biosciences Products
Click to view available options
Quantity:
1 Vial
Description
Detection reagents used in BD™ CBA Human Enhanced Sensitivity Flex Set System consist of two parts, Analyte Specific Detection Reagent (Part A) and Enhanced Sensitivity Detection Reagent (Part B). Enhanced Sensitivity Detection Reagent (Part B) must be used in conjunction with Flex Kit, Master Buffer Kit, flow cytometer and FCAP Array™ Software.
- Provided as lyophilized cake
- Should not be used with Flex Sets from any other BD CBA System such as soluble protein, cell signalling etc
- For QC testing
- Reactivity: human
- Available in vial
Flow Cytometry
Specifications
Specifications
| Cross Reactivity | Human, Mouse |
| Quantity | 1 Vial |
| Content And Storage | 4°C |
| Form | Lyophilized |
| For Use With (Application) | Flow Cytometry, Immunoassay |
| Product Type | Enhanced sensitivity detection reagent (Part B) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction