Learn More
BD Cytometric Bead Array (CBA) Human IL-1β Enhanced Sensitivity Flex Set (Bead B4)
Catalog No. bdb561509 Shop All BD Biosciences Products
Click to view available options
:
Human IL-1beta flex set; 100 tests
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
  • Capture bead
  • Detection reagent
  • Standard
Lyophilized recombinant Human IL-1β measures soluble analytes present in very low concentrations
  • Set is a bead-based immunoassay capable of measuring human interleukin-1β (IL-1β) in serum, plasma and cell culture supernatant samples
  • Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
  • Includes 100 tests
Flow Cytometry
TRUSTED_SUSTAINABILITY
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.