Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human IL-12p70 Enhanced Sensitivity Flex Set (Bead C6)

Catalog No. bdb561518 Shop All BD Biosciences Products
Click to view available options
Quantity:
100 Tests

Includes

  • Kit contents:
  • Enhanced Sensitivity Capture Bead C6: stored in aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
  • Detection Reagent (Part A): stored in aqueous buffered solution containing BSA and ProClin™ 150
  • Standard: lyophilized in an aqueous buffered solution containing BSA and ProClin 150

Measures human Interleukin-12p70 (IL-12p70) in serum, plasma and cell culture supernatant samples

This kit is used in conjunction with BD™ Cytometric Bead Array (CBA) Human Enhanced Sensitivity Master Buffer Kit, flow cytometer and FCAP Array™ Software and can measue soluble analytes present in very low concentrations.

  • Bead-based immunoassay
  • Reactivity: human
  • Standard assay range: 274 to 200000fg/mL

Flow cytometry, Immunoassay

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Lyophilized in an aqueous buffered solution containing BSA™ and ProClin™ 150. Store at 4°C.
Includes Capture beads, detection reagent, standards
For Use With (Application) Used in conjunction with BD™ cytometric bead array (CBA)
No. of Tests 100
Quantity 100 Tests
For Use With (Equipment) Flow Cytometer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.