Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human Inflammatory Cytokine Kit Standard controls
Shop All BD Biosciences Products
Click to view available options
Quantity:
0.2 mL
1 Vial
Product Type:
Human Inflammatory Cytokine Standards
Human Th1/Th2/Th17 Cytokine Standards
Description
- Lyophilized in aqueous buffered solution containing BSA and ProClin™ 150
- Should be reconstituted and serially diluted before use
Human Inflammatory Cytokine Standards
- Contain mixture of recombinant human Interleukin (IL)-8, IL-1β, IL-6, IL-10, Tumor Necrosis Factor (TNF), and IL-12p70
- Use with BD™ Cytometric Bead Array (CBA) Human Inflammatory Cytokine Kit
- Size: 0.2mL
- Contain mixture of recombinant human Interleukin (IL)-2, IL-4, IL-6, IL-10, IL-17A, Interferon-γ (IFN-γ), and Tumor Necrosis Factor (TNF)
- Use with BD™ Cytometric Bead Array (CBA) Human Th1/Th2/TH17 Cytokine Kit
- Size: 1 vial
Flow Cytometry
Specifications
Specifications
| Cross Reactivity | Human |
| Includes | Mixture of recombinant Human Interleukin (IL)-8, IL-1beta, IL-6, IL-10, Tumor Necrosis Factor (TNF) and IL-12p70 |
| Quantity | 0.2 mL |
| Content And Storage | 4°C |
| Form | Lyophilized |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human Inflammatory Cytokine Standards |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction