Learn More
BD Cytometric Bead Array (CBA) Human MIP-1β Flex Set, Bead E4
Catalog No. BDB558288 Shop All BD Biosciences Products
Click to view available options
:
Human MIP-1β; 100 Tests
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
Human MIP-1β; Standard; Human MIP-1β; PE Detection Reagent; Human MIP-1β; Capture Bead E4
For detecting human macrophage inflammatory proteins
The BD CBA Human MIP-1β Flex Set is a bead-based immunoassay capable of measuring human macrophage inflammatory protein-1β (MIP-1β), also known as CCL4, in serum, plasma, and cell culture supernatant samples. Human reactivity was determined by testing samples with the BD CBA Human MIP-1β Flex Set.
  • Bead position: E4
  • Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
  • Standard component is lyophilized
  • When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
  • Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software
TRUSTED_SUSTAINABILITY
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.