Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Mouse IL-13 Enhanced Sensitivity Flex Set (Bead B8)

Catalog No. bdb562273
Click to view available options
Quantity:
100 Tests

Measures mouse Interleukin-13 (IL-13) in cell culture supernatant samples

  • Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
  • Protein concentration of 200,000 fg/ml (when reconstituted in 0.4 ml Assay Diluent)
  • Capable of measuring soluble analytes present in very low concentrations

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-13
Cross Reactivity Mouse
Includes 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized)
Quantity 100 Tests
Sample Type Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Measures Mouse interleukin-13, flow cytometry
Product Type Mouse IL-13 enhanced sensitivity flex set (bead B8)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.