Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Mouse IL-3; Flex Set, Bead A8

Catalog No. bdb558346
Click to view available options
Quantity:
100 Tests

Includes

Mouse IL-3; Standard; Mouse IL-3; PE Detection Reagent; Mouse IL-3; Capture Bead A8

Immunoassay for measuring mouse interleukin-3 in serum and cell culture supernatant samples

The BD CBA Mouse IL-3 Flex Set is a bead-based immunoassay capable of measuring mouse interleukin-3 (IL-3) in serum and cell culture supernatant samples.

  • Reactivity: Mouse
  • Reactivity was determined by testing samples with BD CBA Mouse IL-3 Flex set

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-3
Cross Reactivity Mouse
Includes Mouse IL-3, standard, Mouse IL-3, PE detection reagent, Mouse IL-3, capture bead A8
Quantity 100 Tests
Sample Type Serum, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Mouse IL-3 flex set (bead A8)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.