Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Cytometric Bead Array (CBA) Phospho ERK1/2 (T202/Y204); Flex Set (Bead C4)
Shop All BD Biosciences Products
Click to view available options
Quantity:
100 Tests
Description
Includes
Phospho ERK1/2 (T202/Y204); Standard; Phospho ERK1/2 (T202/Y204); PE Detection Reagent; Phospho ERK1/2 (T202/Y204); Capture Bead C4
- Capable of measuring human, mouse, or rat extracellular signal regulated kinases 1/2 (ERK1/2), a member of the mitogen-activated protein kinase (MAPK) family of kinases, that has been threonine-phosphorylated at Thr-202 and tyrosine-phosphorylated at Tyr-204 in denatured cell lysate samples
- Human and mouse reactivity was determined by testing cell lysates with the BD CBA Phospho ERK1/2 (T202/Y204) Flex Set
- Reactivity: QC Testing: Human; Tested in Development: mouse and rat
- Assay Range: 3.9 to 1000 Units/mL
- Provides sufficient reagents for 100 tests
Order Info
Must be used in conjunction with a BD CBA Cell Signaling Master Buffer Kit (Cat. No. BDB560005, 100 tests, or BDB560006, 500 tests) and a flow cytometer.
Specifications
Specifications
| Antigen | ERK1/2 (T202/Y204) |
| Cross Reactivity | Human, Mouse, Rat |
| Includes | Phospho ERK1/2 (T202/Y204), standard, Phospho ERK1/2 (T202/Y204), PE detection reagent, Phospho ERK1/2 (T202/Y204), capture bead C4 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Phospho ERK1/2 (T202/Y204) flex set (bead C4) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction