Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD D- Luciferin Potassium Salt
Shop All BD Biosciences Products
Click to view available options
Quantity:
10 mg
50 mg
100 mg
250 mg
Description
- Also known as beetle luciferin
- Lyophilized in form of monopotassium salt
- Purified by HPLC to remove dehydroluciferin and L-Luciferin
- Form dehydroluciferin in presence of ultraviolet light and/or moisture
Luminometer

Specifications
Specifications
Formulation | Lyophilized |
Type | D-Luciferin |
Quantity | 100 mg |
Content And Storage | -20°C |
For Use With (Application) | Luminometer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction