Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD D- Luciferin Potassium Salt

Catalog No. bdb556878 Shop All BD Biosciences Products
Click to view available options
Quantity:
10 mg
50 mg
100 mg
250 mg

Synthetically made substrate which can be catalyzed by firefly luciferase

  • Also known as beetle luciferin
  • Lyophilized in form of monopotassium salt
  • Purified by HPLC to remove dehydroluciferin and L-Luciferin
  • Form dehydroluciferin in presence of ultraviolet light and/or moisture

Luminometer

TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized
Type D-Luciferin
Quantity 50 mg
Content And Storage -20°C
For Use With (Application) Luminometer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.