Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Difco™ Casein Digest

Catalog No. DF0116170
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Difco Casein Digest is an animal origin (AO) enzymatic digest of casein with different enzymatic digestion conditions than other casein.

Difco Casein Digest is an animal origin (AO) enzymatic digest of casein with different enzymatic digestion conditions than other casein digests in our portfolio, such as Bacto Casitone and Bacto Tryptone. Difco Casein Digest has a unique nutritional profile of amino acids, nutrients, minerals, nucleotide precursors, peptides, and other metabolites ideal for growth promotion in fermentation applications, especially Escherichia coli.

Features:

  • Unique nutritional profile of amino acids, nutrients, minerals, nucleotide precursors, peptides, and other metabolites

Applications of Difco Casein Digest include:

  • Microbial fermentation
  • Growth promotion, specifically E. Coli

Applications:

Cultivation of heterotrophic microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance LUO, MUO
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Additive Casein Digest
Form Solid
Grade Technical
For Use With (Application) Cultivation of heterotrophic microorganisms
Packaging Type Bottle
Product Line Difco™
Quantity 500 g

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.