Learn More
BD Difco™ Dehydrated Culture Media: Fluid Thioglycollate Medium with Beef Extract
Shop All BD Diagnostic Systems ProductsDescription
- Recommended by the Agricultural Research Service
- Formulation: Beef extract 5g/L, Yeast extract 5g/L, Pancreatic digest of Casein 15g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
Certifications
CE, USP
Specifications
Specifications
| Certifications/Compliance | CE, USP |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Fluid Thioglycollate Medium |
| Product Type | Dehydrated Culture Media |
| Formulation | Beef extract 5g/L, Yeast extract 5g/L, Pancreatic digest of Casein 15g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L |
| For Use With (Application) | Cultivation of microorganisms from normally sterile biological product |
| Packaging Type | Pail |
| Quantity | 10,000 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.