Learn More
BD Difco™ Dehydrated Culture Media: UVM Modified Listeria Enrichment Broth
Shop All BD Diagnostic Systems ProductsSpecifications
Specifications
| Certifications/Compliance | AOAC, CCAM, COMPF, USDA |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Listeria Enrichment Broth |
| Product Type | Microbiology Media |
| Formulation | Pancreatic digest of Casein 5g/L, Proteose Peptone No. 3 5g/L, Beef extract 5g/L, Yeast extract 5g/L, Sodium Chloride 20g/L, Disodium Phosphate 9.6g/L, Monopotassium Phosphate 1.35g/L, Esculin 1g/L, Nalidixic Acid 0.02g/L, Acriflavine HCl 12mg/L |
| For Use With (Application) | For rapidly isolating Listeria monocytogenes |
| Packaging Type | Pail |
| Quantity | 2 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.