Learn More
BD Difco™ Dehydrated Culture Media: Xylose Lysine Deoxycholate (XLD) Agar
Shop All BD Diagnostic Systems ProductsDescription
- Formulation: Xylose 3.5g/L, L-Lysine 5g/L, Lactose 7.5g/L, Saccharose 7.5g/L, Sodium Chloride 5g/L, Yeast extract 3g/L, Phenol red 0.08g/L, Sodium Desoxycholate 2.5g/L, Ferric Ammonium Citrate 0.8g/L, Sodium Thiosulfate 6.8g/L, Agar 13.5g/L
Order Info
Prices are for units of one each unless otherwise specified.
Certifications
AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, ISO, JP, MCM9, SMD, SMWW, USP
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, ISO, JP, MCM9, SMD, SMWW, USP |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | XLD Agar |
| Product Type | Nutrient Agar |
| Formulation | Xylose 3.5g/L, L-Lysine 5g/L, Lactose 7.5g/L, Saccharose 7.5g/L, Sodium Chloride 5g/L, Yeast extract 3g/L, Phenol red 0.08g/L, Sodium Desoxycholate 2.5g/L, Ferric Ammonium Citrate 0.8g/L, Sodium Thiosulfate 6.8g/L, Agar 13.5g/L |
| For Use With (Application) | Isolation and differentiation of enteric pathogens |
| Packaging Type | Pail |
| Quantity | 2 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.