Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Francisella tularensis Antigen and Antisera
Febrile Control Antisera are used in agglutination tests in conjunction with Febrile Antigens as positive controls
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
14-911-128
|
BD
241116 |
Francisella tularensis Antisera |
Each for $776.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
14-911-127
|
BD
241115 |
Francisella tularensis Antigen |
Each for $954.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Aids in diagnosis of certain febrile diseases such as salmonellosis, brucellosis and rickettsial diseases
- Francisella Tularensis Antigen contains 0.5% formaldehyde in place of the phenol
- Febrile Control Antisera are preparations of rabbit antiserum with 30% to 50% glycerin as preservative
- Available in vials of 5mL
For in vitro Diagnostic Use.
Specifications
2°C to 8°C | |
5 mL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title