Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Listeria O Antisera

Catalog No. DF2301501 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Product Type:
Listeria O Antiserum Poly Types 1 and 4
Listeria O Antiserum Type 1
Listeria O Antiserum Type 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
DF2301-50-1 Listeria O Antiserum Type 4
DF2300-50-2 Listeria O Antiserum Type 1
DF2302-50-0 Listeria O Antiserum Poly Types 1 and 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DF2301-50-1 Supplier BD Supplier No. 223011
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used as positive control in identification of Listeria monocytogenes in the slide agglutination test

Listeria monocytogenes is a widespread problem in public health and the food industries.

  • Lyophilized, polyclonal rabbit antisera containing approximately 0.1% sodium azide as a preservative
  • Listeria O Antisera Types 1 and 4 are specific for the respective serotypes of L. monocytogenes while Listeria O Antiserum Poly contains agglutinins for L. monocytogenes serotypes 1 and 4

For in vitro Diagnostic Use.

Certifications

CE marked

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Marked; For in vitro diagnostic use
Content And Storage Refrigerate, store at 2°C to 8°C
Product Type Listeria O Antiserum Type 4
For Use With (Application) Used as positive control in identification of Listeria monocytogenes in the slide agglutination test
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.