Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Leptospira Enrichment EMJH

Catalog No. DF0795731 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
6 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0795-73-1 6 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0795-73-1 Supplier BD Supplier No. 279510
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used with Leptospira Medium Base EMJH in cultivating Leptospira. BD™ Difco™ Leptospira Enrichment EMJH is a lyophilized media additive. The solution contains albumin, polysorbate 80 and additional growth factors for Leptospira.

The basal medium and enrichment are prepared according to the formulations described by Ellinghausen and McCullough, as modified by Johnson and Harris (by replacing rabbit serum medium with polysorbate 80-albumin). Leptospira Medium EMJH is recommended for the clinical isolation of Leptospira from clinical specimens and environmental samples. The Enrichment media contains albumin, polysorbate 80 and additional growth factors for Leptospira.

  • Leptospira Medium Base plus Enrichment shows good culture response to:
    Leptospira interrogans (serotype australis) 23605,
    Leptospira interrogans (serotype canicola) 23470
    Leptospira kirschneri (serotype grippotyphosa) 23604
  • Add 100mL of Enrichment to a solution of 900mL purified water plus 2.3g Medium Base
  • Water content: 89.4%
  • Vapor Pressure (at 20°C): 23.0 hPa (17mm Hg)
  • Soluble in water

In 1886, Adolf Weil described the first recognized leptospiral infections in humans. These cases were caused by Leptospira icterohaemorrhagiae and the disease was subsequently named Weil’s Disease. Leptospirosis is a zoonotic disease, with infection usually resulting from direct or indirect exposure to the urine of leptospiruric animals.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS 11014, SMWW
Color Medium to dark amber
Content And Storage 2°C to 8°C
Additive Leptospira Enrichment
Form Lyophilized
Formulation Albumin, polysorbate 80 and additional growth factors for Leptospira
For Use With (Application) Leptospira medium base EMJH
Packaging Type Bottle
Quantity 6 x 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.