Learn More
Description
- Stable, lyophilized, polyclonal rabbit antisera
- Contains 0.1% sodium azide as preservative
Specifications
Specifications
| Certifications/Compliance | For in vitro diagnostic use |
| Description | Antiserum Group D |
| Product Type | Neisseria Meningitidis Antisera |
| Quantity | 1 mL |
For in vitro diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.