Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Neisseria Meningitidis Antisera

Catalog No. DF2891507 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
1 mL
3 mL
Description:
Antiserum Group A
Antiserum Group B
Antiserum Group C
Antiserum Group D
Antiserum Group W135
Antiserum Group X
Antiserum Group Y
Antiserum Group Z
Antiserum Poly 2, Groups X, Y, Z
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Description
DF2891-50-7 3 mL Antiserum Group Z
DF2230-50-7 1 mL Antiserum Group C
DF2229-50-0 1 mL Antiserum Group B
DF2231-50-6 1 mL Antiserum Group D
DF2228-50-1 1 mL Antiserum Group A
DF2253-50-9 1 mL Antiserum Group W135
DF2880-50-0 1 mL Antiserum Group X
DF2881-50-9 1 mL Antiserum Group Y
DF2910-50-4 3 mL Antiserum Poly 2, Groups X, Y, Z
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. DF2891-50-7 Supplier BD Supplier No. 228911
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

For serotyping Neisseria meningitidis in slide agglutination tests

  • Stable, lyophilized, polyclonal rabbit antisera
  • Contains 0.1% sodium azide as preservative

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance For in vitro diagnostic use
Description Antiserum Group Z
Product Type Neisseria Meningitidis Antisera
Quantity 3 mL

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.