Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Salmonella O QC Antigens

Quick reactive Gold standard antisera-typing panels for Salmonella O antigen

Supplier:  BD 221311/DEL

 View more versions of this product

Catalog No. DF2131-50-7

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

This item is not returnable. View return policy

Description

Description

  • Product type: Antigens and Antisera
  • Groupings: Q through S products
  • Sub type: Quality control antigens
  • Type: Antigens
  • Volume: 1mL
  • Shelf life: 730 days

Certifications

CE marked

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Antigen
Used in the quality control of Salmonella O Antisera Poly B (225351), Factor 7 (228161), Group C1 Factors 6 (240985), and Poly A-I & Vi (240993)
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 730 Days

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.