Learn More
BD Difco™ Salmonella O QC Antigens
Quick reactive Gold standard antisera-typing panels for Salmonella O antigen
Supplier: BD 211753
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
- Product type: Antigens and Antisera
- Groupings: Q through S products
- Sub type: Quality control antigens
- Type: Antigens
- Volume: 1mL
- Shelf life: 730 days
Certifications
CE marked

Specifications
Antigen | |
Used in the quality control of Salmonella O Antisera Poly B (225351), Factor 7 (228161), Group C1 Factors 6 (240985), and Poly A-I & Vi (240993) | |
1 mL |
Safety and Handling
ShelfLife : 730 Days
Your input is important to us. Please complete this form to provide feedback related to the content on this product.