Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Directigen™ EZ Flu A+B Test Kit
Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
30 Tests
Description
Includes
30 Test devices in foil pouches, 4.7mL Reagent E, 2.8mL of each control (A+/B- and B+/A-), 30 BD DispensTube Tubes, 30 BD DispensTube Tips, and liquid controls to allow for easy QC compliance.
- Sensitivity: 86% (Flu A); 80% (Flu B)
- Specificity: 99% (Flu A); 100% (Flu B)
- Sample types: nasopharyngeal washes, nasopharyngeal aspirates and throat swabs
- Two separate chromatographic immunoassays on one device which differentiate between influenza A and B
- Separate reimbursements for Flu A and Flu B enhance financial value
- Optimized mucoid sample processing for improved readability
- Easy-to-read test line
- Compatible with a wide variety of transport media
- Results in 15 minutes
- Internal procedural controls validate proper reagent function and assure that correct test procedure was followed
† CPT reimbursements vary by state. Please refer to the CMS manual or website for the actual reimbursement for your state.

Specifications
Specifications
Clia Complexity | Moderate |
Detectable Analytes | Influenza A and B |
Sample Type | Nasopharyngeal Aspirate, Nasopharyngeal Wash, Throat Swab |
Type | EZ Flu A and B Test Kit |
Includes | 30 Test devices in foil pouches, 4.7mL reagent E, 2.8mL of each control (A+/B- and B+/A-), 30 BD DispensTube Tubes, 30 BD DispensTube Tips and liquid control |
Quantity | 30 Tests |
Test Time | 15 min. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction