Learn More
BD Directigen™ EZ RSV Test Kit
Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
30 Tests
Description
Includes
30 Devices, extraction reagent, swab positive and negative controls, and tubes and tips for specimen extraction and sample delivery into the devices.
- Two-step process, lateral flow test
- Accepts nasopharyngeal washes, aspirates and swabs
- Results within 15 minutes
- Easy-to-read line indicates a positive result
- Includes internal control line
- No interference with commonly used medications
Specifications
Specifications
Clia Complexity | Moderate |
Detectable Analytes | Respiratory Syncytial Virus (RSV) |
Sample Type | Nasal Aspirate, Nasal Swab, Nasal Wash |
Type | EZ RSV Test Kit |
Includes | 30 Devices, extraction reagent, liquid positive and negative control, internal control line and tubes and tips |
Quantity | 30 Tests |
Test Time | 15 min. |