Learn More
BD FITC BrdU Flow Kit
Catalog No. p-4003561 Shop All BD Biosciences Products
Change view
Click to view available options
No. of Tests:
50
Catalog No. No. of Tests
BDB559619 50
1 options
Catalog No. BDB559619
Supplier: BD 559619
Only null left
Add to Cart
Add to Cart
Includes
One 65μL vial fluorochrome-conjugated Anti-BrdU antibody, one 25mL bottle BD Cytofix/Cytoperm™ buffer, two 25mL bottles BD perm/wash™ buffer (10X), one 10mL bottle BD Cytoperm™, DNase; BrdU; 7-AAD; Kit Manual
TRUSTED_SUSTAINABILITY
Specifications
Product Type FITC BrdU Flow Kit
Includes BD Cytofix/Cytoperm™ Buffer; BD Perm/Wash™ Buffer; BD Cytoperm Plus Buffer; FITC Labeled Anti-BrdU Antibody; DNase; BrdU; 7-AAD; Kit Manual
No. of Tests 50
Content And Storage 4°C