Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD H-2Kb:Ig Recombinant Fusion Protein, Mouse
Shop All BD Biosciences Products
Click to view available options
Quantity:
0.25 mg
Description
The H-2K[b]:Ig fusion protein consists of three extracellular major histocompatibility complex (MHC) class I H-2Kb domains that are fused to VH regions of mouse IgG1.
- Format: PE
- Reactivity: Ms
- Isotype: Mouse IgG1, Λ

Specifications
Specifications
Concentration | 0.5mg/mL |
For Use With (Application) | ELISA, Flow Cytometry |
Formulation | Aqueous Buffered solution containing 0.09% Sodium Azide |
Name | Mouse H-2Kb:Ig Recombinant Fusion Protein |
pH Range | 7.2 |
Purification Method | Affinity Chromatography, SDS-PAGE |
Quantity | 0.25 mg |
Storage Requirements | 4°C |
Recombinant | Recombinant |
Form | Solution |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction