Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Hoechst 33342 Solution

Catalog No. bdb561908
Click to view available options
Quantity:
1 mg

Bioimaging certified reagent for fluorescent staining of DNA and nuclei in live or fixed cells

Hoechst 33342 is a bisbenzimidazole dye with high specificity for binding to double-stranded DNA (preferentially binds to A-T base pairs). This dye is useful to label double-stranded DNA and thus to visualize nuclei.

  • Can be excited at ∽355nm by UV light source (UV laser beam or mercury arc-lamp)
  • Emits blue fluorescence light around emission maximum at 461 nm when bound to DNA
  • Ribonuclease treatment is not needed to avoid nonspecific RNA staining
  • Concentration: 1mg/mL
  • Size: 1mg

Bioimaging/Immunofluorescence

TRUSTED_SUSTAINABILITY

Specifications

Product Type Hoechst 33342 solution
Quantity 1 mg
For Use With (Application) Fluorescent Staining, Bioimaging, Immunofluorescence (IF)
Content And Storage 4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.