Learn More
Description
- This stain is excited by the Red laser (with an excitation maximum of 759nm), and has a fluorescence emission maximum of 780nm.
Specifications
Specifications
| Quantity | 200 μg |
| For Use With (Application) |
|
| Storage Requirements | -80°C |
| Stain Type | Fixable Viability Stain 780 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.