Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Horizon Brilliant™ Violet 480 Strepavidin

Catalog No. bdb564876
Click to view available options
Quantity:
0.1 mg

Streptavidin has very high binding affinity for biotin, which is conjugated to the antibody

BD Horizon BV480 Streptavidin is a useful detection reagent for labeling biotinylated primary or secondary antibodies for detection of antigens by immunofluorescence.
Streptavidin was conjugated to BD Horizon BV480 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye has been exclusively developed by BD Biosciences as an optimal dye for imaging applications. BD Horizon BV480 has an Ex Max at 436nm and Em Max at 477nm. The use of a mounting reagent (eg, ProLong™ Gold) is highly recommended to maximize the photostability of BV480.
For confocal microscopy systems, a 440nm laser is the optimal excitation source with optimal emission collection centered at 477nm. BV480 fluorescence can also be excited well by 405nm and 458 nm lasers.
For epifluorescence microscopes with broad spectrum excitation sources, the recommended excitation and emission filters are 445/20nm and 485/20nm bandpass filters, respectively. For specific multicolor imaging applications, the exact filter configurations should be optimized by the end user.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.1 mg/mL
Quantity 0.1 mg
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Content And Storage Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.