Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BV480 Mouse Anti-Human CD20

Catalog No. p-6909047 Shop All BD Biosciences Products
Click to view available options
Quantity:
100 Tests
25 Tests

CD20 is expressed on pre-B-cells, resting and activated B cells, and follicular dendritic cells, but not plasma cells. BD Horizon™ BV480 Mouse Anti-Human CD20 is observed in low levels on a small subset of normal circulating T lymphocytes. The CD20 molecule is involved in the regulation of B-cell activation.

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
TRUSTED_SUSTAINABILITY

Specifications

Product Type BV480 Mouse Anti-Human CD20
Quantity 25 Tests
For Use With (Application) Flow Cytometry
Cross Reactivity QC Testing: Human; Tested in Development: Rhesus, Cynomolgus, Baboon
Form BV480
Sample Volume 5 μL
Content And Storage Store undiluted at 4°C
Gene ID (Entrez) 931
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.