Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BV480 Mouse Anti-Ki-67
Shop All BD Biosciences Products
Click to view available options
Quantity:
100 Tests
25 Tests
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510

Specifications
Specifications
Product Type | BV480 Mouse Anti-Ki-67 |
Quantity | 25 Tests |
For Use With (Application) | Intracellular Staining (Flow Cytometry) |
Cross Reactivity | QC Testing: Human; Tested in Development: Mouse; Reported Reactivity: Rat, Rhesus |
Form | BV480 |
Sample Volume | 5 μL |
Content And Storage | Store undiluted at 4°C |
Storage Buffer | Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction