Description
- Reacts with and covalently binds to cell-surface and intracellular amines
- Permeable plasma cell membranes, such as those present in necrotic cells, allow intracellular diffusion of dye and covalent binding to higher overall concentrations of amines than in non-permeable live cells
- Necrotic cells present in a typical in vitro assay label with higher levels of dye increases their fluorescence intensity 10-20 fold over viable cells
- Labeled cells can be fixed with formaldehyde for downstream decontamination, freezing and/or permeabilization and subsequent intracellular staining while maintaining stable viability stain fluorescence
- Excited by Red laser (with an excitation maximum of 649nm) and has fluorescence emission maximum of 660nm
Flow cytometry, Intracellular staining
Specifications
Specifications
| Type | Fixable Viability Stain |