Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human Definitive and Pancreatic Endoderm Analysis Kit

Catalog No. BDB562496 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562496 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This kit contains harmful, toxic and highly flammable components. Handle with care in accordance with instructions on the package insert.
Catalog No. BDB562496 Supplier BD Supplier No. 562496

missing translation for 'validMainframeMsg'

This kit contains harmful, toxic and highly flammable components. Handle with care in accordance with instructions on the package insert.

Includes

Alexa Fluor™ 488 Mouse anti-Human Pax-6, 25 Tests; PE Mouse anti-PDX-1, 25 Tests; PE Mouse anti-Human FoxA2, 25 Tests; PerCP-Cy™5.5 Mouse anti-Sox2, 25 Tests; PerCP-Cy5.5 Mouse anti-Human Sox17, 25 Tests; Alexa Fluor 647 Mouse anti-Human Nanog, 25 Tests; Perm Buffer III, 50mL; Fixation Buffer, 50mL

Kit contains a combination of mouse monoclonal antibody conjugates

Cell Populations Identified:

  • Pluripotent stem cells
  • Definitive endoderm
  • Pancreatic endoderm
  • Ectodermal contaminants
  • Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
For Use With (Application) Flow Cytometry
Quantity 25 Tests
Reactivity Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.