Learn More
Description
The OptEIA™ set for human interleukin-10 (IL-10) contains components necessary to develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant human IL-10 in serum, plasma, and cell culture supernatants.
- Capture antibody: anti-human IL-10
- Detection antibody: biotinylated anti-human IL-10
- Enzyme reagent: streptavidin-horseradish peroxidase conjugate (SAv-HRP)
- Standard: lyophilized recombinant human IL-10
- Standard range: 7.8 - 500 pg/mL
ELISA
Specifications
Specifications
| Concentration | 7.8 to 500pg/mL |
| Product Type | Human IL-10 ELISA set |
| Sufficient For | 20 96-well Plates |
| For Use With (Application) | To develop ELISA for natural or recombinant Human IL-10 in Serum, Plasma and cell culture supernatants |
| Kit Contents | anti-Human IL-10 (capture) Antibody, Biotinylated anti-Human IL-10 (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human IL-10 (standard), Streptavidin-HRP Conjugate |
| Quantity | 20 Plates |
| Storage Requirements | 2°C to 8°C |
For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.