Learn More
Description
Includes
- Instrument Setup Bead F1
- Instrument Setup Bead F9
- Instrument Setup Bead A1
- Instrument Setup Bead A9
- Assay Diluent
- Detection Reagent Diluent
- F Wash Buffer (1X)
- PE Instrument Setup Bead F1
- PE Positive Control Detector
- Capture Bead Diluent
Contains all of the supporting reagents necessary to perform an assay using CBA Human Immunoglobulin Flex Set.
- Provides sufficient reagents for the quantitative analysis of 100 samples and 10 instrument setup procedures
- Buffers and instrument setup reagents provided, have been optimized for analysis of immunoglobulin in serum samples.
Specifications
Specifications
| Quantity | 100 Tests |
| Product Type | Cytometric Bead Array (CBA) Assay |
| Includes | 2 bottles 40mL assay diluent, 1 bottle 5mL capture bead diluent, 1 bottle 5mL detection reagent diluent, 1 bottle 260mL wash buffer, 1 vial 0.25mL instrument setup bead A1, 1 vial 0.25mL instrument setup bead A9, 1 vial, 1.0 mL instrument setup bead F1 |
| No. of Tests | 100.0 |
| Content And Storage | 2° to 8°C |
| Validated Application | RUO |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.