Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human intracellular CytoKine-2 Positive Control Cells, HiCK-2

Catalog No. bdb555062 Shop All BD Biosciences Products
Click to view available options
Quantity:
1 mL

Includes

5x106 activated human PBMCs (20 test samples at 2.5 x 105 cells/test) fixed and suspended in 1mL (10% dimethylsulfoxide and 90% fetal bovine serum)

Contains fixed, non-permeabilized human lymphoid cells

  • Includes cells that express detectable levels of intracellular IL-3, IL-4, IL-10,IL-13 and GM-CSF
  • Prepared by stimulating human PBMCs in the presence of a protein transport inhibitor
  • Cells harvested were fixed and then stored in 1mL of 10% dimethylsulfoxide and 90% fetal bovine serum at -80°
  • Concentration: 5x106 cells/mL

Intracellular staining, Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Color Light Brown
Format Frozen
Reactivity Human
Concentration 5 x 106 cells/mL
Detects IL-3, IL-4, IL-10,IL-13 and GM-CSF
For Use With (Application) Intracellular staining
Kit Contents ≈5 x 106 Activated Human PBMCs (20 test samples at 2.5 x 105 cells/test) fixed and suspended in 1mL
Quantity 1 mL
Content And Storage -80°C

For research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.