Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Human TGF-β1 ELISA Set
Shop All BD Biosciences Products
Click to view available options
Quantity:
20 Plates
Description
Includes
Capture Antibody: Anti-human TGF-β1, Detection Antibody: Biotinylated anti-human TGF-β1, Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP), Standard: Recombinant human TGF-β1 (lyophilized), Instruction/Analysis Certificate.
- Flexible assay design
- Calibrated against purified recombinant human TGF-β1

Specifications
Specifications
Concentration | 62.5 to 4000pg/mL |
Product Type | Human TGF-beta1 ELISA set |
Sufficient For | 20 x 96-well Plates |
For Use With (Application) | To develop ELISA for natural or recombinant Human TGF-beta1 in Serum, Plasma and cell culture supernatants |
Kit Contents | anti-Human TGF-beta1 monoclonal (capture) Antibody, Biotinylated anti-Human TGF-beta1 monoclonal (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human TGF-beta1 (standard), Streptavidin-HRP Conjugate |
Quantity | 20 Plates |
Storage Requirements | 2°C to 8°C |
Research use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction