Description
- For needleless collection of blood
- Holder with preattached multiple sample, male adapter
- Compatible with female luer connections or IV ports designed for luer access
- Allows specimens to be collected into an evacuated tube
- Has blue color-coded threads to provide easy differentiation from other holder-based products
- Provides the security of a threaded locking luer connection, replacing luer slip devices
- Compatible with luer-activated ports for closed, direct urine sampling found in some Foley catheters (such as the Bard™ E-Z™ Lock Sampling Port)
- Sterile; latex-free
Specifications
Specifications
| Sterility | Sterile |
| For Use With (Application) | For needle-less collection of Blood |
| Product Type | Blood Collection Access Device |
| Connectors | Male Luer |
| For Use With (Equipment) | Luer activated ports for closed, direct Urine sampling found in some Foley catheters |
| Latex Free | Yes |
Non-returnable medical device