Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD MitoScreen Flow Cytometry Mitochondrial Membrane Potential Detection Kit

Catalog No. bdb551302 Shop All BD Biosciences Products
Click to view available options
Quantity:
100 Tests

Includes

JC-1 Dye and Assay Buffer

Contains JC-1, which has been optimized for use in flow cytometry, and 10 assay buffer

  • Contains 100% JC-1 (5,5',6,6'-tetrachloro-1,1',3,3'-tetraethylbenzimidazolcarbocyanine iodide) dye, an irritant
  • JC-1, typically excited using 488nm line of an argon ion laser while the monomeric form emits at 527nm and J-aggregates at 590nm

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RUO
Content And Storage 2° to 8°C
Includes JC-1 dye and assay buffer: 100% JC-1 (5,5',6,6'-tetrachloro-1,1',3,3'-tetraethylbenzimidazolcarbocyanine iodide) dye, an irritant
No. of Tests 100
Quantity 100 Tests
For Use With (Equipment) Flow Cytometer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.