Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Mouse IFN-γ ELISA Set

Catalog No. bdb555138 Shop All BD Biosciences Products
Click to view available options
Quantity:
20 Plates

Includes

  • Capture Antibody: Anti-mouse IFN-Γ monoclonal antibody
  • Detection Antibody: Biotinylated anti-mouse IFN-Γ monoclonal antibody
  • Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP)
  • Standards: Recombinant mouse IFN-Γ, lyophilized
  • Instruction / Analysis Certificate

For immunoassay protocols

The OptEIA set for mouse interferon-Γ (IFN-Γ) contains the components necessary to develop enzyme-linked immunosorbent assays (ELISA).

  • Reactivity: Mouse
  • For natural or recombinant mouse IFN-Γ in serum, plasma, and cell culture supernatants

Enzyme Linked Immunosorbent Assay

TRUSTED_SUSTAINABILITY

Specifications

Product Type Mouse IFN-gamma ELISA set
Sufficient For 20 96-well Plates
For Use With (Application) To develop ELISA for natural or recombinant Mouse IFN-g in Serum, Plasma and cell culture supernatants
Kit Contents anti-Mouse IFN-gamma monoclonal (capture) antibody, Biotinylated anti-Mouse IFN-gamma monoclona (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse IFN-g (standard), Streptavidin-HRP Conjugate
Quantity 20 Plates
Storage Requirements 2°C to 8°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.