Learn More
BD Mouse Intracellular Cytokine Staining Starter Kit
Catalog No. bdb559311 Shop All BD Biosciences Products
Click to view available options
Quantity:
25 Tests
Includes
BD PharMingen Stain Buffer (FBS); Cytofix/Cytoperm Buffer; Perm/Wash Buffer; PE-conjugated anti-human or mouse IL-2; PE-conjugated anti-human or mouse IFN-g; PE-conjugated anti-human or mouse TNF-a; PE-conjugated isotype control cocktail; Purified blocking antibody cocktail; Leukocyte Activation Cocktail; MiCK-1 or HiCK-1 Cytokine Positive Control Cells; Staining Kit Manual
Serves as an introduction to the intracellular staining technique
  • Contains the essential reagents for the activation of a mouse spleen cell population and their subsequent intracellular staining to detect IL-2, IFN-γ-, and TNF-a-producing cells
TRUSTED_SUSTAINABILITY
Specifications
Detects IL-2, IFN-g- and TNF-a
Quantity 25 Tests
Content And Storage 4°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.