Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Native Protein

Catalog No. bdb558618 Shop All BD Biosciences Products
Click to view available options
Quantity:
10 Tests

Useful as a standard in a sandwich ELISA to measure mouse C3a protein levels

Purified Mouse C3a protein is a native protein isolated from activated mouse serum. Anaphylatoxin C3a is a bioactive cleavage product released from plasma component C3 during complement activation and is involved in mediation of a variety of cellular immune responses, as well as being a potent pro-inflammatory agent. The release of this cleavage product is a reliable indicator of in vivo or in vitro complement activation.

  • Storage buffer: Lyophilized in an aqueous buffered solution containing BSA and ProClin 150
  • Isolated and purified by affinity chromatography

ELISA

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA
Formulation Lyophilized in an aqueous Buffered solution containing BSA and ProClin 150
Name Mouse C3a Native Protein
pH Range 6.5
Preparation Method In vivo or in vitro complement
Purification Method Affinity Chromatography
Quantity 10 Tests
Source Mouse Serum
Storage Requirements 4°C
Cross Reactivity Mouse
Recombinant Recombinant
Form Lyophilized
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.