Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD OptEIA™ Reagent Set B

Catalog No. bdb550534
Click to view available options
Quantity:
1 Set

Includes

Wash Concentrate 20X, substrate reagent A, substrate reagent B, stop solution, assay diluent, coating buffer

Sufficient for twenty 96-well plates

Enzyme-linked immunosorbent assay

TRUSTED_SUSTAINABILITY

Specifications

Product Type OptEIA™ reagent set B
Quantity 1 Set
For Use With (Application) ELISA
Content And Storage 2°C to 8°C
Kit Contents 250mL Coating buffer pH 9.5, 1L Assay diluent, 1L Wash concentrate 20X, 125mL Substrate reagent A, 125mL Reagent B, 125mL Stop solution
Sufficient For 20 96-well Plates
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.