Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD PBX Calcium Assay Kit

Catalog No. BDB640175 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
10 Preps
100 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB640175 10 Preps
BDB640177 100 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB640175 Supplier BD Supplier No. 640175
Add to Cart
Add to Cart

Includes

Calcium Indicator, 10X PBX Signal Enhancer

For homogeneous measurement of intracellular calcium changes

  • Involves one dye addition step
  • Does not require washing
  • Allows easy implementation in highthroughput environment
  • Probenecid, a standard inhibitor of nonspecific anion transport, is not required for dye-loading
  • Can improve retention of the dye in cells

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
For Use With (Application) For homogeneous measurement of intracellular calcium changes
Includes 1 vial calcium indicator,10mL PBX signal enhancer (10X, lyophilized), 90mL calcium assay buffer 1X
Quantity 10 Preps
Type Calcium Assay Kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.