Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Persist™ Skin Prep Swabs

Catalog No. 22265644 Shop All BD Diagnostic Systems Products
Click to view available options
Product Type:
Single Skin Prep Swab
This item is not returnable. View return policy
This item is not returnable. View return policy

Fast and easy one-step application

  • 10% PVP-I and 70% alcohol
  • Provides up to seven days of antimicrobial efficacy under an occlusive dressing
  • Lays the foundation for IV catheter start
  • Dries in less than a munute

TRUSTED_SUSTAINABILITY

Specifications

Product Type Single Skin Prep Swab
Composition 70% Alcohol and 10% Povidone Iodine
For Use With (Application) Kills microorganisms immediately on contact
Latex Free Yes
Sterility Non-sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.