Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD DRAQ5™

Catalog No. BDB564903 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564903 50 μL
BDB564902 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB564903 Supplier BD Supplier No. 564903
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to stain nucleic acids in live or fixed cells. BD Pharmingen™ DRAQ5™ is also used for cell cycle analysis by DNA content, nuclear visualization, or discrimination of nucleated cells from debris or enucleated cells.

  • Far-red fluorescent DNA dye
  • Cell permeable
  • Used to stain nucleic acids in live or fixed cells
  • Has an excitation wavelength maxima of 598/646nm
TRUSTED_SUSTAINABILITY

Specifications

Color Far-red
Concentration 5 mM
Content And Storage Store undiluted at 4°C
For Use With (Application) Bioimaging, Immunofluorescence, Intracellular Staining (Flow Cytometry)
Dye Type DNA dye
Quantity 50 μL
Reactivity Tested in Development: Human, Mouse; Reported Reactivity: Rat
Excitation Wavelength 598/646 nm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.